Learn More
Trajan Scientific Syringe Gas tight GT 250uL 1/EA
SDP
Catalog No. 5000494947
Encompass_Preferred
250 L syringe with gas tight plunger plunger stop and fixed Luer Lock
Catalog No. 50-004-94947 Supplier Trajan Scientific Supplier No. 006232
250 L syringe with gas tight plunger plunger stop and fixed Luer Lock