Learn More
Trajan Scientific Syringe Gas tight GT 250uL 1/EA
SDP
Supplier:  Trajan Scientific 006720
Encompass_Preferred
250 L fixed needle CTC syringe with gas tight plunger and 5 1 cm 0 72 mm OD LC tipped needle
Catalog No. 50-004-94315