Learn More
Trajan Scientific Syringe Gas tight GT 250uL 1/EA
Supplier: Trajan Scientific 006862
250 L fixed needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 syringe with gas tight plunger and 5 7 cm 0 63 mm OD cone tipped needle