Learn More
Trajan Scientific Syringe Gas tight GT 25uL 1/EA
SDP
Supplier:  Trajan Scientific 003864
Encompass_Preferred
25 L fixed needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 syringe with gas tight plunger and 8 5 cm 0 63 mm OD cone tipped needle
Catalog No. 50-004-94334