Learn More
Trajan Scientific Syringe Gas tight GT 25uL 1/EA
SDP
Supplier:  Trajan Scientific 003988
Encompass_Preferred
25 L removable needle CTC Analytics and Thermo Scientific syringe with gas tight plunger and 5 cm 0 47 mm OD side hole needle
Catalog No. 50-004-94761