Learn More
Trajan Scientific Syringe Gas tight GT 25uL 1/EA
SDP
Catalog No. 5000494094
Encompass_Preferred
25 L removable needle CTC MS syringe with gas tight plunger
Catalog No. 50-004-94094 Supplier Trajan Scientific Supplier No. 003770
25 L removable needle CTC MS syringe with gas tight plunger