Learn More
Trajan Scientific Syringe Gas tight GT 500uL 1/EA
SDP
Catalog No. 5000494875
Encompass_Preferred
500 L syringe with gas tight plunger and 1/4-28 UNF termination
Catalog No. 50-004-94875 Supplier Trajan Scientific Supplier No. 007995
500 L syringe with gas tight plunger and 1/4-28 UNF termination