Learn More
Trajan Scientific Syringe Gas tight GT 500uL 1/EA
SDP
Catalog No. 5000494113
Encompass_Preferred
500 L syringe with gas tight plunger and M10 termination
Catalog No. 50-004-94113 Supplier Trajan Scientific Supplier No. 007660
500 L syringe with gas tight plunger and M10 termination