Learn More
Trajan Scientific Syringe Gas tight GT 500uL 1/EA
SDP
Catalog No. 5000494819
Encompass_Preferred
500 L fixed needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 syringe with gas tight plunger and 5 7 cm 0 63 mm OD side hole needle
Catalog No. 50-004-94819 Supplier Trajan Scientific Supplier No. 007890
500 L fixed needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 syringe with gas tight plunger and 5 7 cm 0 63 mm OD side hole needle