Learn More
Trajan Scientific Syringe Gas tight GT 500uL 1/EA
SDP
Supplier:  Trajan Scientific 007866
Encompass_Preferred
500 L fixed needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 syringe with gas tight plunger and 5 7 cm 0 47 mm OD cone tipped needle
Catalog No. 50-004-94849