Learn More
Trajan Scientific Syringe Gas tight GT 500uL 1/EA
SDP
Catalog No. 5000494783
Encompass_Preferred
500 L fixed needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 syringe with gas tight plunger and 8 5 cm 0 63 mm OD cone tipped needle
Catalog No. 50-004-94783 Supplier Trajan Scientific Supplier No. 007864
500 L fixed needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 syringe with gas tight plunger and 8 5 cm 0 63 mm OD cone tipped needle