Learn More
Trajan Scientific Syringe Gas tight GT 50uL 1/EA
SDP
Catalog No. 5000494642
Encompass_Preferred
50 L removable needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 MS syringe with gas tight plunger
Catalog No. 50-004-94642 Supplier Trajan Scientific Supplier No. 004790
50 L removable needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 MS syringe with gas tight plunger