Learn More
Trajan Scientific Syringe Gas tight GT 50uL 1/EA
SDP
Catalog No. 5000493799
Encompass_Preferred
51 L syringe with gas tight plunger and fixed Luer Lock
Catalog No. 50-004-93799 Supplier Trajan Scientific Supplier No. 004230CAL
51 L syringe with gas tight plunger and fixed Luer Lock