Learn More
Trajan Scientific Syringe Gas tight GT 50uL 1/EA
SDP
Supplier:  Trajan Scientific 004279
Encompass_Preferred
50 L syringe with gas tight plunger and removable needle push-pull valve with 5 cm 0 63 mm OD cone tipped needle
Catalog No. 50-004-93954