Learn More
Trajan Scientific Syringe Gas tight GT 50uL 1/EA
SDP
Catalog No. 5000494843
Encompass_Preferred
50 L syringe with gas tight plunger and fixed Luer tip
Catalog No. 50-004-94843 Supplier Trajan Scientific Supplier No. 004229
50 L syringe with gas tight plunger and fixed Luer tip