Learn More
Trajan Scientific Syringe Gas tight GT 50uL 1/EA
SDP
Supplier:  Trajan Scientific 004312
Encompass_Preferred
50 L removable needle syringe with gas tight plunger and 5 1 cm 0 72 mm OD LC tipped needle
Catalog No. 50-004-94342