Learn More
Trajan Scientific Syringe Gas tight GT 50uL 1/EA
SDP
Supplier:  Trajan Scientific 004668
Encompass_Preferred
50 L fixed needle Agilent syringe with gas tight plunger and 4 2 cm 0 63/0 47 mm OD cone tipped dual gauge needle
Catalog No. 50-004-94828