Learn More
Trajan Scientific Syringe Gas tight GT 50uL 1/EA
SDP
Catalog No. 5000494486
Encompass_Preferred
50 L syringe with gas tight plunger plunger stop and fixed Luer Lock
Catalog No. 50-004-94486 Supplier Trajan Scientific Supplier No. 004232
50 L syringe with gas tight plunger plunger stop and fixed Luer Lock