Learn More
Trajan Scientific Syringe Gas tight GT 50uL 1/EA
SDP
Catalog No. 5000494489
Encompass_Preferred
50 L removable needle CTC MS syringe with gas tight plunger
Catalog No. 50-004-94489 Supplier Trajan Scientific Supplier No. 004770
50 L removable needle CTC MS syringe with gas tight plunger