Learn More
Trajan Scientific Syringe Gas tight GT 50uL 1/EA
SDP
Catalog No. 5000494495
Encompass_Preferred
50 L removable needle MS syringe with gas tight plunger
Catalog No. 50-004-94495 Supplier Trajan Scientific Supplier No. 004750
50 L removable needle MS syringe with gas tight plunger