Learn More
Trajan Scientific Syringe Gas tight GT 50uL 1/EA
SDP
Catalog No. 5000494833
Encompass_Preferred
50 L fixed needle CTC syringe with gas tight plunger and 5 1 cm 0 72 mm OD LC tipped needle
Catalog No. 50-004-94833 Supplier Trajan Scientific Supplier No. 004810
50 L fixed needle CTC syringe with gas tight plunger and 5 1 cm 0 72 mm OD LC tipped needle