Learn More
Trajan Scientific Syringe Gas tight GT 50uL 1/EA
SDP
Catalog No. 5000494834
Encompass_Preferred
50 L syringe with gas tight plunger and 1/4-28 UNF termination
Catalog No. 50-004-94834 Supplier Trajan Scientific Supplier No. 004995
50 L syringe with gas tight plunger and 1/4-28 UNF termination