Learn More
Trajan Scientific Syringe Gas tight GT 5mL 1/EA
SDP
Catalog No. 5000494702
Encompass_Preferred
5 mL headspace soil gas tight syringe with gas tight plunger and push-button valve with Luer Lock termination removable side hole Luer probe and 5 cm 0 63 mm OD bevel tipped needle
Catalog No. 50-004-94702 Supplier Trajan Scientific Supplier No. 008775
5 mL headspace soil gas tight syringe with gas tight plunger and push-button valve with Luer Lock termination removable side hole Luer probe and 5 cm 0 63 mm OD bevel tipped needle