Learn More
Chemglass Life Sciences Syringe, Micro, Gas-Tight, 500uL capacity, PTFE Luer Lock
SDP
Catalog No. CG307422
Encompass_Preferred
Syringe, Micro, Gas-Tight, 500uL capacity, PTFE Luer Lock. Syringes are designed for use in both liquid and gas chromatography. Due to the all-PTFE tip, they can also handle highly corrosive or Ultra-pure liquids. With the exacting tolerances held in the manufacture of these syringes, all components are interchangeable within each size range.PLEASE NOTE: PTFE LUER TIP OR LUER LOCK SYRINGES ARE NOT SUPPLIED WITH NEEDLES. FOR NEEDLES SEE CG-3075.
Catalog No. CG307422 Supplier Chemglass Life Sciences Supplier No. CG307422
Syringe, Micro, Gas-Tight, 500uL capacity, PTFE Luer Lock. Syringes are designed for use in both liquid and gas chromatography. Due to the all-PTFE tip, they can also handle highly corrosive or Ultra-pure liquids. With the exacting tolerances held in the manufacture of these syringes, all components are interchangeable within each size range.PLEASE NOTE: PTFE LUER TIP OR LUER LOCK SYRINGES ARE NOT SUPPLIED WITH NEEDLES. FOR NEEDLES SEE CG-3075.