Learn More
Braintree Scientific SyringeHeater for Sm Syringes spiral,kit w/ribbon heater pad
SDP
Catalog No. NC0892836
Encompass
Syringe Heater Kit Heating device for syringes that require temperature-controlled dispensing Heats syringe to approximately 185C Compatiblity Notes: Heating syringes over 100C with BS-300, BS-500, BS-8000 & BS-9000 model syringe pumps requires an affordable customized pump. Call for details. Digitally set the heater setpoint Temperature sensor in heating pad Setpoint retained in memory Controller will heat the syringe and hold the set temperature using an on/off or PID control algorithm Multiple syringe heater pads can be attached to the control unit Heated syringe can be mounted on a syringe pump Syringe Heater Kit includes Control-Unit power cable and Primary-Syringe-Heater-Pad **Sold for research purposes only and not for human use**
Catalog No. NC0892836 Supplier Braintree Scientific Supplier No. BSSYRHSM
Syringe Heater Kit Heating device for syringes that require temperature-controlled dispensing Heats syringe to approximately 185C Compatiblity Notes: Heating syringes over 100C with BS-300, BS-500, BS-8000 & BS-9000 model syringe pumps requires an affordable customized pump. Call for details. Digitally set the heater setpoint Temperature sensor in heating pad Setpoint retained in memory Controller will heat the syringe and hold the set temperature using an on/off or PID control algorithm Multiple syringe heater pads can be attached to the control unit Heated syringe can be mounted on a syringe pump Syringe Heater Kit includes Control-Unit power cable and Primary-Syringe-Heater-Pad **Sold for research purposes only and not for human use**