Learn More
GA International Syringe Identification Labels for Thermal-Transfer Printers 1.5" x 1" + 2.5" (tail) / 38.1mm x 25.4mm + 63.5mm (tail) Rectangle White + Transparent From -80°C to +100°C (-112°F to +212°F)
Catalog No. 5000571802
Encompass_Preferred
SyringeTAG™ labels are ideal for identifying syringes used in hospitals, clinics, laboratories, and other medical facilities. The two-sided print area is attached to a clear tail that wraps around the syringe. The flagging printed area displays vital information such as patient ID, drug, dosage, 1 or 2-D barcodes, graphic images, and more. SyringeTAG labels fit most syringe sizes (1 ml, 2 ml, 5 ml, and 10 ml) and have a service temperature range from as -80°C (-112°F) to 100°C (212°F). Available in roll format. Intended for use with all standard brands of thermal-transfer printers or to be written on with permanent marker. For the most durable print, we recommend our RR-class ribbons.
Catalog No. 50-005-71802 Supplier GA International Supplier No. SYRB1RSNOTC11.5WH
SyringeTAG™ labels are ideal for identifying syringes used in hospitals, clinics, laboratories, and other medical facilities. The two-sided print area is attached to a clear tail that wraps around the syringe. The flagging printed area displays vital information such as patient ID, drug, dosage, 1 or 2-D barcodes, graphic images, and more. SyringeTAG labels fit most syringe sizes (1 ml, 2 ml, 5 ml, and 10 ml) and have a service temperature range from as -80°C (-112°F) to 100°C (212°F). Available in roll format. Intended for use with all standard brands of thermal-transfer printers or to be written on with permanent marker. For the most durable print, we recommend our RR-class ribbons.