Learn More
Chemglass Life Sciences Syringe, Luer Lock, 20cc Capacity, 1cc Subdivisions
SDP
Catalog No. CG307005
Encompass_Preferred
Syringe, Luer Lock, 20cc Capacity, 1cc Subdivisions. Borosilicate glass syringe has interchangeable barrels and plungers. Colored plunger ends identify the capacity of the syringe. Flanges are flat on two sides to prevent syringe from rolling. All sizes of syringes are equipped with a metal luer lock and permanently fused graduations. For needles see CG-3075.
Catalog No. CG307005 Supplier Chemglass Life Sciences Supplier No. CG307005
Syringe, Luer Lock, 20cc Capacity, 1cc Subdivisions. Borosilicate glass syringe has interchangeable barrels and plungers. Colored plunger ends identify the capacity of the syringe. Flanges are flat on two sides to prevent syringe from rolling. All sizes of syringes are equipped with a metal luer lock and permanently fused graduations. For needles see CG-3075.