Learn More
Med Vet International Dynarex Syringe - Luer Lock, 20 Cc, 500/Case
Catalog No. 502831611
Encompass_Preferred
Dynarex Syringes without Needles can be used with any size needle with a luer slip or luer lock connection. These Syringes are designed to provide accurate and effective medication dosing in a variety of medical settings. These Syringes feature transparent barrels with bold, easy-to-read markings for precise measurement. Dynarex Syringes Without Needle have smooth-gliding integrated plungers to provide consistent pressure for improved comfort. Dynarex Syringes without Needles are high-quality syringes designed to provide accurate and effective medication dosing. Each Syringe features a transparent barrel with bold, easy-to-read numbering and either a luer lock or luer slip connection. The smooth-gliding plunger provides seamless and consistent pressure for greater patient comfort.
Catalog No. 50-283-1611 Supplier Med Vet International Supplier No. 6991
Dynarex Syringes without Needles can be used with any size needle with a luer slip or luer lock connection. These Syringes are designed to provide accurate and effective medication dosing in a variety of medical settings. These Syringes feature transparent barrels with bold, easy-to-read markings for precise measurement. Dynarex Syringes Without Needle have smooth-gliding integrated plungers to provide consistent pressure for improved comfort. Dynarex Syringes without Needles are high-quality syringes designed to provide accurate and effective medication dosing. Each Syringe features a transparent barrel with bold, easy-to-read numbering and either a luer lock or luer slip connection. The smooth-gliding plunger provides seamless and consistent pressure for greater patient comfort.