Learn More
WELCH MATERIALS INC Welchrom Syringe Filter, MCE, 13mm, 0.45μm, Prefilter, Double layers,100pcs/pk,10pk/cs.
Catalog No. NC2052776
Encompass_Preferred
Syringe Filter, MCE, 13mm, 0.45μm, Prefilter, Double layers,100pcs/pk,10pk/cs.
Catalog No. NC2052776 Supplier WELCH MATERIALS INC Supplier No. S13MCE045B10
Syringe Filter, MCE, 13mm, 0.45μm, Prefilter, Double layers,100pcs/pk,10pk/cs.