Learn More
Trajan Scientific Syringe Metal 100uL Removable Needle 1/EA
SDP
Catalog No. 5000494428
Encompass_Preferred
100 L removable needle syringe with 7 cm 0 5 mm OD bevel tipped needle
Catalog No. 50-004-94428 Supplier Trajan Scientific Supplier No. 005055
100 L removable needle syringe with 7 cm 0 5 mm OD bevel tipped needle