Learn More
Trajan Scientific Syringe Metal 10uL Fixed Needle 1/EA
SDP
Supplier:  Trajan Scientific 002400CAL
Encompass_Preferred
10 L fixed needle guided plunger syringe with 5 cm 0 47 mm OD bevel tipped needle calibrated
Catalog No. 50-004-94548