Learn More
Trajan Scientific Syringe Metal 10uL Fixed Needle 1/EA
SDP
Catalog No. 5000494550
Encompass_Preferred
10 L fixed needle CTC Analytics Thermo Scientific AS3000 AS2000 Varian 8400 SCION CP-8410 and CP-8400 syringe with 5 cm 0 47 mm OD cone tipped needle calibrated
Catalog No. 50-004-94550 Supplier Trajan Scientific Supplier No. 0029801CAL
10 L fixed needle CTC Analytics Thermo Scientific AS3000 AS2000 Varian 8400 SCION CP-8410 and CP-8400 syringe with 5 cm 0 47 mm OD cone tipped needle calibrated