Learn More
Trajan Scientific Syringe Metal 10uL Fixed Needle 1/EA
SDP
Supplier:  Trajan Scientific 002300
Encompass_Preferred
10 L fixed needle syringe with Superflex plunger and 5 1 cm 0 72 mm OD LC tipped needle
Catalog No. 50-004-93977