Learn More
Trajan Scientific Syringe Metal 10uL Fixed Needle 1/EA
SDP
Supplier:  Trajan Scientific 002105
Encompass_Preferred
10 L fixed needle syringe with flexible plunger and 5 cm 0 47 mm OD cone tipped needle
Catalog No. 50-004-93963