Learn More
Trajan Scientific Syringe Metal 10uL Fixed Needle 1/EA
SDP
Catalog No. 5000494709
Encompass_Preferred
10 L fixed needle syringe with 5 cm 0 47 mm OD cone tipped needle
Catalog No. 50-004-94709 Supplier Trajan Scientific Supplier No. 002005
10 L fixed needle syringe with 5 cm 0 47 mm OD cone tipped needle