Learn More
Trajan Scientific Syringe Metal 10uL Fixed Needle 1/EA
SDP
Supplier:  Trajan Scientific 002992
Encompass_Preferred
10 L fixed needle CTC autosampler and Thermo TriPlus syringe with 80 mm 26 gauge 0 47 mm OD cone tipped needle
Catalog No. 50-004-94263