Learn More
Trajan Scientific Syringe Metal 10uL Fixed Needle 1/EA
SDP
Catalog No. 5000494493
Encompass_Preferred
10 L fixed needle CTC Analytics Thermo Scientific AS3000 AS2000 Varian 8400 SCION CP-8410 and CP-8400 syringe with 5 cm 0 5 mm OD cone tipped needle
Catalog No. 50-004-94493 Supplier Trajan Scientific Supplier No. 0029671
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10 L fixed needle CTC Analytics Thermo Scientific AS3000 AS2000 Varian 8400 SCION CP-8410 and CP-8400 syringe with 5 cm 0 5 mm OD cone tipped needle