Learn More
Trajan Scientific Syringe Metal 10uL Fixed Needle 1/EA
SDP
Catalog No. 5000494474
Encompass_Preferred
10 L fixed needle CTC syringe with 5 1 cm 0 72 mm OD LC tipped needle
Catalog No. 50-004-94474 Supplier Trajan Scientific Supplier No. 002710
10 L fixed needle CTC syringe with 5 1 cm 0 72 mm OD LC tipped needle