Learn More
Trajan Scientific Syringe Metal 10uL Fixed Needle 1/EA
SDP
Supplier:  Trajan Scientific 002867
Encompass_Preferred
10 L fixed needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 syringe with 8 5 cm 0 47 mm OD cone tipped needle
Catalog No. 50-004-94725