Learn More
Trajan Scientific Syringe Metal 10uL Fixed Needle 25/PK
SDP
Catalog No. 5000494544
Encompass_Preferred
10 L fixed needle syringe with 5 cm 0 47 mm OD bevel tipped needle PK25
Catalog No. 50-004-94544 Supplier Trajan Scientific Supplier No. 002035
10 L fixed needle syringe with 5 cm 0 47 mm OD bevel tipped needle PK25