Learn More
Trajan Scientific Syringe Metal 10uL Fixed Needle 25/PK
SDP
Supplier:  Trajan Scientific 002813
Encompass_Preferred
10 L fixed needle Agilent syringe with 4 2 cm 0 63 mm OD cone tipped needle PK25
Catalog No. 50-004-94813