Learn More
Trajan Scientific Syringe Metal 10uL Fixed Needle 6/PK
SDP
Supplier:  Trajan Scientific 0029861
Encompass_Preferred
10 L fixed needle CTC Analytics Thermo Scientific AS3000 AS2000 Varian 8400 SCION CP-8410 and CP-8400 syringe with 5 cm 0 47 mm OD cone tipped needle PK6
Catalog No. 50-004-94329