Learn More
Trajan Scientific Syringe Metal 10uL Fixed Needle 6/PK
SDP
Catalog No. 5000494660
Encompass_Preferred
10 L fixed needle syringe with 5 1 cm 0 72 mm OD LC tipped needle PK6
Catalog No. 50-004-94660 Supplier Trajan Scientific Supplier No. 002315
10 L fixed needle syringe with 5 1 cm 0 72 mm OD LC tipped needle PK6