Learn More
Trajan Scientific Syringe Metal 10uL Fixed Needle 6/PK
SDP
Supplier:  Trajan Scientific 002130
Encompass_Preferred
10 L fixed needle syringe with flexible plunger and 5 cm 0 47 mm OD bevel tipped needle PK6
Catalog No. 50-004-94948