Learn More
Trajan Scientific Syringe Metal 10uL Removable Needle 1/EA
SDP
Supplier:  Trajan Scientific 002805
Encompass_Preferred
10 L removable needle Agilent syringe with 4 2 cm 0 47 mm OD cone tipped needle
Catalog No. 50-004-94245