Learn More
Trajan Scientific Syringe Metal 10uL Removable Needle 1/EA
SDP
Catalog No. 5000494525
Encompass_Preferred
10 L removable needle CTC Analytics and Thermo Scientific syringe with 5 cm 0 63 mm OD cone tipped needle
Catalog No. 50-004-94525 Supplier Trajan Scientific Supplier No. 002984
10 L removable needle CTC Analytics and Thermo Scientific syringe with 5 cm 0 63 mm OD cone tipped needle