Learn More
Trajan Scientific Syringe Metal 10uL Removable Needle 1/EA
SDP
Catalog No. 5000494762
Encompass_Preferred
10 L removable needle CTC RTC and Thermo RSH autosampler syringe with 5 7 cm 23 gauge 0 63 mm OD cone tipped needle
Catalog No. 50-004-94762 Supplier Trajan Scientific Supplier No. 002871
10 L removable needle CTC RTC and Thermo RSH autosampler syringe with 5 7 cm 23 gauge 0 63 mm OD cone tipped needle