Learn More
Trajan Scientific Syringe Metal 10uL Removable Needle 1/EA
SDP
Supplier:  Trajan Scientific 0029931
Encompass_Preferred
10 L removable needle CTC Analytics Thermo Scientific AS3000 AS2000 Varian 8400 SCION CP-8410 and CP-8400 syringe with 8 cm 0 47 mm OD cone tipped needle
Catalog No. 50-004-94485