Learn More
Trajan Scientific Syringe Metal 10uL Removable Needle 1/EA
SDP
Catalog No. 5000494455
Encompass_Preferred
10 L removable needle syringe with 5 cm 0 47 mm OD bevel tipped needle
Catalog No. 50-004-94455 Supplier Trajan Scientific Supplier No. 002050
10 L removable needle syringe with 5 cm 0 47 mm OD bevel tipped needle