Learn More
Trajan Scientific Syringe Metal 10uL Removable Needle 1/EA
SDP
Supplier:  Trajan Scientific 002993
Encompass_Preferred
10 L removable needle CTC autosampler and Thermo TriPlus syringe with 80 mm 26 gauge 0 47 mm OD cone tipped needle
Catalog No. 50-004-93952